Learn More
National Safety Apparel™ Custom Arc H2O Pant Flo. Orange/Red Tall
SDP
Catalog No. 170012292 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2292 5X-Large
17-001-2285 Small
17-001-2286 Medium
17-001-2287 Large
17-001-2288 X-Large
17-001-2289 2X-Large
17-001-2290 3X-Large
17-001-2291 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-2292 Supplier National Safety Apparel™ Supplier No. R50RQ195XT
Specifications
Size 5X-Large
Gender Unisex
Material Polyurethane-coated FR Cotton
Color Fluorescent Orange
Ankle Style Open
Seam Type Welded and Taped
Closure Type Buckle
Disposable Reusable
Arc Flash Rating 19 cal/cm² (Fluorescent Yellow); 24 cal/cm² (Fluorescent Orange)
Certifications/Compliance CAT 2, NFPA 70 E, ANSI/ISEA 107 Class E, CSA Z462, ASTM F2733, ASTM F1891, OSHA 1910.269
Sterility Non-sterile