Learn More
Electron Microscopy Sciences Aurion ImmunoGold Reagents Custom Kit
SDP
Catalog No. 5028286
Encompass
Immunogold reagent kit. Two non-overlapping gold particle sizes, Coupled to: either Goat-anti-Rabbit and Goat-anti-Mouse IgG/IgM antibodies. Affinity purified and cross adsorbed antibodies.
Catalog No. 50-282-86 Supplier Electron Microscopy Sciences Supplier No. 2554005
Immunogold reagent kit. Two non-overlapping gold particle sizes, Coupled to: either Goat-anti-Rabbit and Goat-anti-Mouse IgG/IgM antibodies. Affinity purified and cross adsorbed antibodies.