Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Cyclohexane/Ethyl Acetate 1mL in a 2mL ampoule

Catalog No. 502398643 Shop All Phenomenex Products
Encompass_Preferred

Custom Pesticides Mix 3-Aroclors 200 g/mL in AL0-130141

Catalog No. 50-239-8643 Supplier Phenomenex Inc Supplier No. AL0130141
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Custom Pesticides Mix 3-Aroclors 200 g/mL in AL0-130141

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.