Learn More
StatLab Custom Pre-fill Buffered Formalin for Dermpath

Description
- Conveniently prefilled with 10% Neutral Buffered Formalin for secure tissue fixation.
- Each 40 mL container is filled with 20 mL of NBF
- Each 20 mL container is filled with 10 mL of NBF
- 96 prefills per case
Specifications
Specifications
| Product Type | Pre-filled Formalin Container |
| Closure Included | Yes |
| Graduated | Yes |
| Material | Polypropylene |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Capacity (Metric) | 20 mL |
| Color | Clear |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.