Learn More
StatLab Custom Pre-fill Buffered Formalin for SEP

Description
- Produces an audible click when the Leak-proof seal is properly engaged, helping ensure secure closure and specimen integrity during transport
- Available in StatLabs proprietary HistoPak carrier for easy distribution or in bulk packaging
- 95 kPa certified for Leak-resistant transport in compliance with DOT and IATA standards
- Containers are filled to 50% capacity, 7 mL containers are prefilled with 5 mL
- Customization available with laboratory name and logo
- Conveniently prefilled with 10% Neutral Buffered Formalin for quick and efficient tissue fixation.
- Each 40 mL container is filled with 20 mL of NBF
- Each 20 mL container is filled with 10 mL of NBF
- Includes 96 prefills per case
Specifications
Specifications
| Product Type | Pre-filled Formalin Container |
| Closure Included | Yes |
| Graduated | Yes |
| Material | Polypropylene |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Capacity (Metric) | 20 mL |
| Color | Clear |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.