Learn More
StatLab Custom Pre-fill Zinc Buffered Formalin

Description
- Custom prefilled zinc buffered formalin for PRW only
- Especially recommended for use in immunohistochemistry, as zinc formalin preserves more immunoreactivity than standard NBF
- Each 40 mL container is filled with 20 mL of NBF
- 96 prefills per case
Specifications
Specifications
| Product Type | Pre-filled Zinc Formalin Container |
| Closure Included | Yes |
| Graduated | Yes |
| Material | Polypropylene |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Capacity (Metric) | 20 mL |
| Color | Clear |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.