Learn More
Ohiolumex Co Inc High-speed robust fiber-optic oxygen probe, REDFLASH technology, T90 < 0.5 s gas, T90 < 3 s water, 0-500 hPa oxygen, 1 bar max, 10 cm length, 3 mm diameter, 2 m cable, stainless steel, ST-connector
SDP
Catalog No. NC3397541
Encompass
High-speed robust fiber-optic oxygen probe, REDFLASH technology, T90 < 0.5 s gas, T90 < 3 s water, 0-500 hPa oxygen, 1 bar max, 10 cm length, 3 mm diameter, 2 m cable, stainless steel, ST-connector
Catalog No. NC3397541 Supplier Ohiolumex Co Inc Supplier No. PSOXROBHSTL200CL2
High-speed robust fiber-optic oxygen probe, REDFLASH technology, T90 < 0.5 s gas, T90 < 3 s water, 0-500 hPa oxygen, 1 bar max, 10 cm length, 3 mm diameter, 2 m cable, stainless steel, ST-connector