Learn More
Analytical Sales & Services Custom SS HPLC Fittings Kit for 1/16″ OD Tubing
Catalog No. 5000804482
Encompass
Advantage Custom SS HPLC Fittings Kit Contains (4) ZDV Unions (2) Packages of Nuts & (2) Packages of Ferrules
Qty/Pkg: EACH
Catalog No. 50-008-04482 Supplier Analytical Sales & Services Supplier No. 414315
Advantage Custom SS HPLC Fittings Kit Contains (4) ZDV Unions (2) Packages of Nuts & (2) Packages of Ferrules
Qty/Pkg: EACH