Learn More
Perkin Elmer US LLC Custom Standard QC 1A, 500 mL
Catalog No. 502113669
Encompass_Preferred
Al, Cu @5 µg/mL Mg @ 10 µg/mL, Zn @ 20 µg/mL Fe, Pb @ 25 µg/mL, Ca @ 50 µg/mL B @ 250 µg/mL, P @1000 µg/mL K @ 2500 µg/mL, Na @ 5000 µg/mL Matrix: 3% HNO3 (v/v) Volume: 500 mL
Catalog No. 50-211-3669 Supplier Perkin Elmer US LLC Supplier No. N9308342
Al, Cu @5 µg/mL Mg @ 10 µg/mL, Zn @ 20 µg/mL Fe, Pb @ 25 µg/mL, Ca @ 50 µg/mL B @ 250 µg/mL, P @1000 µg/mL K @ 2500 µg/mL, Na @ 5000 µg/mL Matrix: 3% HNO3 (v/v) Volume: 500 mL