Learn More
Applied Biosystems™ Custom TaqMan™ SNP Genotyping Assays
Description
Applied Biosystems TaqMan SNP Genotyping Assays use TaqMan 5'-nuclease chemistry to amplify and detect specific polymorphisms in purified genomic DNA samples. Each assay enables genotyping of individuals for a single nucleotide polymorphism (SNP) and consists of two sequence-specific primers and two TaqMan minor groove binder (MGB) probes with non-fluorescent quenchers (NFQ). Assays contain one probe labeled with VIC or Cyanine 5 dye to detect the Allele 1 sequence; the second probe is labeled with FAM or ABY dye to detect the Allele 2 sequence.
Custom TaqMan SNP Genotyping Assays can be easily designed at no extra charge by submitting target sequences confidentially to our secure Custom Assay Design Tool.
Benefits:
- Proven—gold-standard TaqMan chemistry and robust assay designs deliver accurate, reproducible, and reliable results
- Easy—convenient single-tube format and simple workflow provide an easy path to trusted results; no optimization required
- Flexible—obtain assay designs for any SNP in the human genome using our secure Custom Assay Design Tool, at no extra charge
- Tested—all custom human assays (VIC/FAM SKUs only) are subjected to a functional test on 20 unique DNA samples. Assays ordered with non-human or Cyanine 5/ABY SKUs are not functionally tested.
- Multiplexing ready—TaqMan assays are now available with with VIC/FAM or Cyanine 5/ABY formats to support simultaneous detection of two sets of SNPs (two pairs of alleles).
Approximate ship time
4–6 days in North America and 6–10 days in Europe (VIC/FAM); 4-10 days in North America, 6-14 days in Europe (Cyanine 5/ABY)
The free Custom Assay Design Tool can generate assays for the detection of SNPs, MNPs, and indels of up to six bases. These custom assays are designed, synthesized, formulated, optimized, and quality control tested.
TaqMan SNP Genotyping Assays require only three reaction components for PCR: purified genomic DNA (1–20 ng), the assay solution, and TaqMan Genotyping Master Mix (singleplex) or TaqPath ProAmp Master Mix (multiplex) (sold separately).
All assay designs are the product of our industry-leading bioinformatics pipeline, optimized over the course of more than a decade by leveraging manufacturing and assay performance data. TaqMan Assays have been cited in over 40,000 publications and are backed by more than 350 patents.
Recommended master mix (sold separately): TaqMan Genotyping Master Mix (Cat. No. 4371355) or TaqPath ProAmp Master Mix (Cat. No. A30865)
Specifications
Specifications
| Quantity | L (2400 reactions), Made to Order |
| Product Line | TaqMan |
| For Use With (Equipment) | 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ Absolute Q Digital PCR System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex ProFlex PCR System*, VeritiPro*, SimpliAmp*, MiniAmp*, Automated Thermal Cycler** If a thermal cycler is used for PCR amplification, the optional pre-read and the post-read must be performed separately on a real-time PCR system in order to detect and record fluorescent signals. |
| Shipping Condition | Room Temperature |
| Sample Type | Non-human |
| Concentration | 80X |
| Content And Storage | 1 tube containing a 40X (S and M sizes) or 80X (L size) mix of pre-formulated assay (2 probes and 2 primers). Store at -15 to -25°C. |
| No. of Reactions | 12,000 (384-well), 2400 (96-well) |
Frequently Asked Questions (FAQs)
We recommend using ~1-20 ng of good-quality gDNA per well. The gDNA should have an A260/A280 ratio of ~1.7-1.9. It is also important to try to add the same amount of gDNA to every well.
You can use our Custom Assay Design Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/real-time-pcr-assays/snp-genotyping-taqman-assays/custom-snp-assays.html?ICID=ta-lm-custom%20taqman%20snp%20assay-Single%20Tube%20Custom%20TaqMan%C2%AE%20SNP%20Genotyping%20Assays) to submit your SNP sequence for an assay design. Alternatively, you can also use Primer Express Software to design an assay. The DNA sequence needs to have at least 30 bases on each side of the SNP site. The more sequence you provide, the greater chance you have of the tool creating a passing design. The ideal sequence length is ~ 200 bases on either side of the SNP site. For the best-performing assay, we recommend prescreening the sequence for other SNPs, repeats, and regions of low complexity. These variations should be masked prior to submitting the sequence. This will prevent the design tool from designing the primers or probe over those regions, which could reduce the efficiency of the assay if they were used. For more details on how to screen and mask the sequence, please follow the guidelines here (https://tools.thermofisher.com/content/sfs/manuals/cms_042307.pdf).
We recommend diluting the 40X and 80X TaqMan SNP Genotyping Assays to a 20X working stock with 1X TE buffer. The 1X TE buffer should be 10 mM Tris-HCl, 1 mM EDTA, pH 8.0, and be made using DNase-free, sterile-filtered water.
You can utilize the Applied Biosystems TaqMan Multiplex Interaction Check (MIC) tool. The MIC tool uses bioinformatics to identify potential interactions between primers, probes, and amplicons that could negatively impact multiplex qPCR performance. The tool was developed and validated using data from approximately 200 multiplex panels, combining computational analysis with wet-lab testing. By flagging assay combinations that may be prone to false positives or reduced sensitivity, MIC tool helps researchers design multiplex panels with greater confidence and reduce time spent on trial-and-error optimization.risk and observed multiplex performance.
You can multiplex up to two Applied Biosystems TaqMan SNP Genotyping Assays to interrogate two sets of alleles. The first assay will utilize FAM/VIC dyes, while the second assay will utilize ABY/Cyanine 5 dyes.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.