Learn More
Neogen Diagnostics Custom TMB Substrate, 200 ML, HRP, Microwell, Colorimetric
Catalog No. NC2215566
Encompass_Preferred
Custom TMB Substrate is a one bottle stabilized chromogenic substrate for use with horseradish peroxidase (HRP) immunoassays. It contains both 3,3,5,5 tetramethylbenzidine (TMB) and hydrogen peroxide (H2O2). Custom TMB Substrate develops a deep blue color in the presence of peroxidase labelled conjugate. This substrate has been optimized for use with microwell assays, especially ELISA. Custom TMB is not for use with assays requiring a precipitating substrate. Stable for a minimum of 36-months when stored at 2-8C.
This item is not returnable. View return policy
Catalog No. NC2215566 Supplier Neogen Diagnostics Supplier No. 309175
Custom TMB Substrate is a one bottle stabilized chromogenic substrate for use with horseradish peroxidase (HRP) immunoassays. It contains both 3,3,5,5 tetramethylbenzidine (TMB) and hydrogen peroxide (H2O2). Custom TMB Substrate develops a deep blue color in the presence of peroxidase labelled conjugate. This substrate has been optimized for use with microwell assays, especially ELISA. Custom TMB is not for use with assays requiring a precipitating substrate. Stable for a minimum of 36-months when stored at 2-8C.