Learn More
Electron Microscopy Sciences Aurion ImmunoGold Custom Ultra Small Kit
SDP
Supplier:  Electron Microscopy Sciences 2555005
Encompass_Preferred
Immunogold reagent kit. Customize the Ultra Small Kitsby choosing two vials of your Ultra Small Reagent. Also supplied with the kit is 30ml BSA-c and 2x30ml of R-Gent SE-LM.
Catalog No. 50-282-89