Learn More
MSC HexArmor Cut/Puncture Sleeve, SuperFabric, ANSI Cut A7, Large, 1 ea
Catalog No. 502886345
Encompass
HexArmor cut and puncture-resistant sleeves are designed for industrial applications, providing superior protection for the forearm. Made with advanced SuperFabric material, these sleeves offer exceptional cut and puncture resistance.
  • Innovative industrial puncture protection from wires, metal, wood, and glass over entire forearm
  • Won't fall down like knit sleeves
  • Lighter and cooler than knit or leather alternatives
  • Lasts 13 times longer than competitor's sleeves
  • Spandex wrist insert with thumbhole
  • Launderable
  • Lightweight
  • Low-linting
Catalog No. 50-288-6345 Supplier MSC Supplier No. 72052616
HexArmor cut and puncture-resistant sleeves are designed for industrial applications, providing superior protection for the forearm. Made with advanced SuperFabric material, these sleeves offer exceptional cut and puncture resistance.
  • Innovative industrial puncture protection from wires, metal, wood, and glass over entire forearm
  • Won't fall down like knit sleeves
  • Lighter and cooler than knit or leather alternatives
  • Lasts 13 times longer than competitor's sleeves
  • Spandex wrist insert with thumbhole
  • Launderable
  • Lightweight
  • Low-linting