Learn More
Grainger Cut Off Blade 1/16 Carbide Tip
Catalog No. 502766539
Encompass_Preferred
The Grainger 16T501 is a 16-ton bottle jack made of steel with a lift range of 10-15/16 inches to 17-1/8 inches. It has a built-in overload valve to prevent over-extension and can lift equipment, dies, molds, and more. This hydraulic jack has a saddle diameter of 5-1/8 inches and an extended height of 17-1/8 inches. It meets ASME PALD standards and comes with a one year warranty from Grainger.
Catalog No. 50-276-6539 Supplier Grainger Supplier No. 16T501
The Grainger 16T501 is a 16-ton bottle jack made of steel with a lift range of 10-15/16 inches to 17-1/8 inches. It has a built-in overload valve to prevent over-extension and can lift equipment, dies, molds, and more. This hydraulic jack has a saddle diameter of 5-1/8 inches and an extended height of 17-1/8 inches. It meets ASME PALD standards and comes with a one year warranty from Grainger.