Learn More
Grainger Cut Off Blade 1/8 Carbide Tip
Catalog No. 502766570
Encompass_Preferred
The Cut-Off Blade, 1/8" Carbide Tip, 16T522 from Grainger is a 1/8" thick cut-off blade with a carbide tip. It has a 1" arbor hole and is 6" in diameter. This abrasive cut-off wheel is rated for 15,300 RPM max and is designed for cutting steel, stainless steel, aluminum and other metals. It should be used with a saw guard for safety. This is a high quality, durable cut-off wheel made in the USA by Weiler Corporation.
Catalog No. 50-276-6570 Supplier Grainger Supplier No. 16T522
The Cut-Off Blade, 1/8" Carbide Tip, 16T522 from Grainger is a 1/8" thick cut-off blade with a carbide tip. It has a 1" arbor hole and is 6" in diameter. This abrasive cut-off wheel is rated for 15,300 RPM max and is designed for cutting steel, stainless steel, aluminum and other metals. It should be used with a saw guard for safety. This is a high quality, durable cut-off wheel made in the USA by Weiler Corporation.