Learn More
Grainger Cut Off Blade 1/8 Carbide Tip
Supplier:  Grainger 16T523
Encompass_Preferred
The Grainger 1/8" Carbide Tip Cut-Off Blade is a circular saw blade designed for cutting metal. It features a 1/8" thickness with a 1" arbor hole and measures 4-1/2" in diameter. The blade has 16 teeth made of carbide tipped steel for durability when cutting metal materials. This cut-off blade can be used with circular saws, chop saws, and portable band saws when precise cuts in metal are required. The thin kerf design allows for efficient, clean cuts.
Catalog No. 50-276-4878