Learn More
Grainger Cut Off Blade 1/8 Carbide Tip
Supplier:  Grainger 16T516
Encompass_Preferred
The Micro 100 Cut-Off Blade is a 1-8" carbide-tipped blade designed for cutting metal, plastic, and wood. It has 16 teeth and fits standard cut-off holders. The blade is made of M2 high speed steel with TiCN coating for hardness, durability and heat resistance. It can cut steel up to 45 HRC hardness. The blade produces minimal burrs and provides clean cuts. It has a thickness of 0.032" and comes in packs of 5. The blade is manufactured in the USA by Micro 100, a leading maker of precision cutting tools.
Catalog No. 50-276-6458