Learn More
Grainger Cut Off Blade 1/8 Carbide Tip
Catalog No. 502766454
Encompass_Preferred
The Grainger 16T506 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for a variety of cleaning applications. This 8" x 4" hand pad has an open web construction that allows it to conform to surfaces. It is made of high-quality fibers that are durable and long-lasting. The pad is designed for scrubbing and cleaning while being gentle on surfaces. It can be used wet or dry on a variety of materials including stainless steel, aluminum, plastic, glass, tile, porcelain, fiberglass and painted wood. This hand pad from 3M and Grainger is a useful cleaning tool for commercial and industrial applications.
Catalog No. 50-276-6454 Supplier Grainger Supplier No. 16T506
The Grainger 16T506 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for a variety of cleaning applications. This 8" x 4" hand pad has an open web construction that allows it to conform to surfaces. It is made of high-quality fibers that are durable and long-lasting. The pad is designed for scrubbing and cleaning while being gentle on surfaces. It can be used wet or dry on a variety of materials including stainless steel, aluminum, plastic, glass, tile, porcelain, fiberglass and painted wood. This hand pad from 3M and Grainger is a useful cleaning tool for commercial and industrial applications.