Learn More
Grainger Cut Off Blade 5/32 Carbide Tip
Catalog No. 502766150
Encompass_Preferred
The Grainger 16T525 is a Dayton 1/3 HP 115/230V Dual Voltage Farm Duty Electric Motor. It is a general purpose motor with a 1/3 horsepower rating and operates on either 115 or 230 volt power. The motor has a 1075 RPM operating speed with a 56C frame size. It features permanent split capacitor construction and is thermally protected. Applications include driving pumps, fans, and shop tools. The motor dimensions are 7-1/2" diameter x 8-3/4" long and it weighs 15 pounds.
Catalog No. 50-276-6150 Supplier Grainger Supplier No. 16T525
The Grainger 16T525 is a Dayton 1/3 HP 115/230V Dual Voltage Farm Duty Electric Motor. It is a general purpose motor with a 1/3 horsepower rating and operates on either 115 or 230 volt power. The motor has a 1075 RPM operating speed with a 56C frame size. It features permanent split capacitor construction and is thermally protected. Applications include driving pumps, fans, and shop tools. The motor dimensions are 7-1/2" diameter x 8-3/4" long and it weighs 15 pounds.