Learn More
Grainger Cut Off Blade 5/64 Carbide Tip
Catalog No. 502767405
Encompass_Preferred
The Grainger 16T503 is a 16 ton capacity hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-1/8 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It is designed for portability with its compact size and enclosed hydraulic system. The jack's lift range makes it suitable for use with light and medium duty automotive and truck lifting applications. Its hydraulic power system enables smooth lifting and controlled lowering of loads up to 16 tons with minimal effort. This high quality hydraulic jack is made in the USA and is backed by a 12 month warranty from Hein-Werner.
Catalog No. 50-276-7405 Supplier Grainger Supplier No. 16T503
The Grainger 16T503 is a 16 ton capacity hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-1/8 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It is designed for portability with its compact size and enclosed hydraulic system. The jack's lift range makes it suitable for use with light and medium duty automotive and truck lifting applications. Its hydraulic power system enables smooth lifting and controlled lowering of loads up to 16 tons with minimal effort. This high quality hydraulic jack is made in the USA and is backed by a 12 month warranty from Hein-Werner.