Learn More
Grainger Cut-Off GDM 1516L003PF15D PR1535 10 pcs.
Catalog No. 502768539
Encompass_Preferred
The Grainger 170FR9 is a 9-inch, 170 CFM axial duct fan made by Continental Fan Manufacturing Inc. It features a direct drive motor with permanently lubricated ball bearings and is thermally protected. This duct fan has a galvanized steel housing and is designed for general ventilation and exhaust applications. With a maximum temperature rating of 104°F, it requires 115V power and draws up to 1.1 amps while operating at 1075 RPM. The model 170FR9 weighs approximately 15 lbs and has a sound rating of 67 dBA. It can be mounted vertically or horizontally for inline air flow using the pre-punched mounting holes.
Catalog No. 50-276-8539 Supplier Grainger Supplier No. 170FR9
The Grainger 170FR9 is a 9-inch, 170 CFM axial duct fan made by Continental Fan Manufacturing Inc. It features a direct drive motor with permanently lubricated ball bearings and is thermally protected. This duct fan has a galvanized steel housing and is designed for general ventilation and exhaust applications. With a maximum temperature rating of 104°F, it requires 115V power and draws up to 1.1 amps while operating at 1075 RPM. The model 170FR9 weighs approximately 15 lbs and has a sound rating of 67 dBA. It can be mounted vertically or horizontally for inline air flow using the pre-punched mounting holes.