Learn More
Grainger Cut-Off GDM 1516N003PF PR1225 10 pcs.
Catalog No. 502765269
Encompass_Preferred
The Grainger 170FP9 is a 9-inch, 170 CFM axial duct fan made by Continental Fan Manufacturing Inc. It features a direct drive motor with permanently lubricated ball bearings and is thermally protected. This duct fan has a galvanized steel housing and is designed for general ventilation and exhaust applications. With a maximum temperature rating of 104°F, it requires 115V power and draws up to 1.1 amps while operating at 1075 RPM. The model 170FP9 adds 970 cubic feet per minute of air flow to HVAC systems or ventilation projects.
Catalog No. 50-276-5269 Supplier Grainger Supplier No. 170FP9
The Grainger 170FP9 is a 9-inch, 170 CFM axial duct fan made by Continental Fan Manufacturing Inc. It features a direct drive motor with permanently lubricated ball bearings and is thermally protected. This duct fan has a galvanized steel housing and is designed for general ventilation and exhaust applications. With a maximum temperature rating of 104°F, it requires 115V power and draws up to 1.1 amps while operating at 1075 RPM. The model 170FP9 adds 970 cubic feet per minute of air flow to HVAC systems or ventilation projects.