Learn More
Grainger Cut-Off GDM 1516N003PF PR1535 10 pcs.
Catalog No. 502765447
Encompass_Preferred
The Grainger 170FR4 is a 4-ton floor jack with a lifting range of 5-1/4 to 18-1/4 inches. It has a steel frame and saddle and uses a hydraulic lifting mechanism to raise loads up to 8,000 lbs. This heavy duty jack has swivel casters for easy maneuverability. It meets ASME PALD standards and comes with a one year warranty from Grainger. The floor jack weighs 85 lbs and measures 28-1/2 x 12-1/4 x 6-3/4 inches.
Catalog No. 50-276-5447 Supplier Grainger Supplier No. 170FR4
Add to Cart
Add to Cart
The Grainger 170FR4 is a 4-ton floor jack with a lifting range of 5-1/4 to 18-1/4 inches. It has a steel frame and saddle and uses a hydraulic lifting mechanism to raise loads up to 8,000 lbs. This heavy duty jack has swivel casters for easy maneuverability. It meets ASME PALD standards and comes with a one year warranty from Grainger. The floor jack weighs 85 lbs and measures 28-1/2 x 12-1/4 x 6-3/4 inches.