Learn More
Grainger Cut-Off GDM 1516R003PF15D PR1225 10 pcs.
Catalog No. 502766548
Encompass_Preferred
The 170FR0 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is a 6 liter stainless steel cylinder that contains the Novec 1230 clean agent. This agent is intended for use on Class A, B, and C fires. The extinguisher has a discharge range of 10-15 feet and can operate between -4°F to 120°F. It features a wall-mounting bracket and discharge hose with a pistol grip nozzle. The extinguisher is UL rated 2-A:40-B:C and meets NFPA 2010 requirements. It does not contain any ozone-depleting substances.
Catalog No. 50-276-6548 Supplier Grainger Supplier No. 170FR0
The 170FR0 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is a 6 liter stainless steel cylinder that contains the Novec 1230 clean agent. This agent is intended for use on Class A, B, and C fires. The extinguisher has a discharge range of 10-15 feet and can operate between -4°F to 120°F. It features a wall-mounting bracket and discharge hose with a pistol grip nozzle. The extinguisher is UL rated 2-A:40-B:C and meets NFPA 2010 requirements. It does not contain any ozone-depleting substances.