Learn More
Grainger Cut-Off GDM 1516R015PF15D PR1225 10 pcs.
Catalog No. 502764679
Encompass_Preferred
The 170FW3 is a 3-phase, 460V AC, 60Hz, 3HP, 1750 RPM, TEFC electric motor manufactured by Baldor-Reliance. It features a cast iron frame, thermally protected, Class F insulation, and 1.15 service factor. The motor dimensions are 9-7/16" diameter x 15-3/16" long with a C-face mounting. It has a NEMA design B torque characteristic and meets NEMA MG1 Part 31 standards. The motor is suitable for industrial applications requiring reliable 3HP electric power.
Catalog No. 50-276-4679 Supplier Grainger Supplier No. 170FW3
The 170FW3 is a 3-phase, 460V AC, 60Hz, 3HP, 1750 RPM, TEFC electric motor manufactured by Baldor-Reliance. It features a cast iron frame, thermally protected, Class F insulation, and 1.15 service factor. The motor dimensions are 9-7/16" diameter x 15-3/16" long with a C-face mounting. It has a NEMA design B torque characteristic and meets NEMA MG1 Part 31 standards. The motor is suitable for industrial applications requiring reliable 3HP electric power.