Learn More
Grainger Cut-Off GDM 2020L003PF15D PR1535 10 pcs.
Catalog No. 502765774
Encompass_Preferred
The Grainger 170FT1 is a Dayton 1/3 HP Thermoplastic Submersible Sump Pump that features a thermoplastic construction and can pump up to 3900 GPH at 0 feet of head. It has a 1/3 HP motor, is 115V, has a vertical float switch, and 1-1/2" NPT discharge. The pump is designed for pumping clear water with temperatures up to 86°F in sump pits up to 8 feet deep. It can handle solids up to 1/2" in diameter. The pump measures 9-3/4" high x 5-3/4" diameter and weighs 11 pounds. It comes with a 10' cord and is UL listed.
Catalog No. 50-276-5774 Supplier Grainger Supplier No. 170FT1
The Grainger 170FT1 is a Dayton 1/3 HP Thermoplastic Submersible Sump Pump that features a thermoplastic construction and can pump up to 3900 GPH at 0 feet of head. It has a 1/3 HP motor, is 115V, has a vertical float switch, and 1-1/2" NPT discharge. The pump is designed for pumping clear water with temperatures up to 86°F in sump pits up to 8 feet deep. It can handle solids up to 1/2" in diameter. The pump measures 9-3/4" high x 5-3/4" diameter and weighs 11 pounds. It comes with a 10' cord and is UL listed.