Learn More
Grainger Cut-Off GDM 2020N003PF PR1225 10 pcs.
Catalog No. 502766299
Encompass_Preferred
The Grainger 170FN4 is a 4-inch diameter, 3-blade, aluminum propeller for small unmanned aerial vehicles. It has a pitch of 4 inches and is designed for electric motors up to 250 Kv. The propeller is made from durable, lightweight 6061 aluminum alloy and uses a set screw to mount to the motor shaft. With high efficiency airfoils, this propeller provides good thrust for multirotor drones and RC aircraft. The max power is 300 watts and max speed is 11,000 rpm. Overall, this is a high quality aluminum propeller well-suited for hobbyist drones and RC vehicles using electric brushless motors.
Catalog No. 50-276-6299 Supplier Grainger Supplier No. 170FN4
The Grainger 170FN4 is a 4-inch diameter, 3-blade, aluminum propeller for small unmanned aerial vehicles. It has a pitch of 4 inches and is designed for electric motors up to 250 Kv. The propeller is made from durable, lightweight 6061 aluminum alloy and uses a set screw to mount to the motor shaft. With high efficiency airfoils, this propeller provides good thrust for multirotor drones and RC aircraft. The max power is 300 watts and max speed is 11,000 rpm. Overall, this is a high quality aluminum propeller well-suited for hobbyist drones and RC vehicles using electric brushless motors.