Learn More
Grainger Cut-Off GDM 2020N015PF PR1225 10 pcs.
Catalog No. 502764836
Encompass_Preferred
The 170FW0 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Fan Cooled, TEFC Electric Motor manufactured by Baldor-Reliance. It has a 1.15 service factor, Class B insulation, and a rigid base. The motor is totally enclosed and fan cooled with a cast iron frame and end brackets. It operates on single phase 115V power with a full load amp rating of 9.2A. The motor shaft is 5/8" in diameter and 1-7/16" long. It rotates at 1725 RPM and outputs 0.37 kW of mechanical power. The motor is built for continuous duty operation.
Catalog No. 50-276-4836 Supplier Grainger Supplier No. 170FW0
The 170FW0 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Fan Cooled, TEFC Electric Motor manufactured by Baldor-Reliance. It has a 1.15 service factor, Class B insulation, and a rigid base. The motor is totally enclosed and fan cooled with a cast iron frame and end brackets. It operates on single phase 115V power with a full load amp rating of 9.2A. The motor shaft is 5/8" in diameter and 1-7/16" long. It rotates at 1725 RPM and outputs 0.37 kW of mechanical power. The motor is built for continuous duty operation.