Learn More
Grainger Cut-Off GDM 2020N020PM PR1535 10 pcs.
Catalog No. 502764763
Encompass_Preferred
The WIDIA GTD Thread Forming Tap is a high speed steel tap designed for forming threads in ductile materials like aluminum, brass, mild steels, and some plastics. It has a TiN coating for increased wear resistance and lubricity. This is a M10 x 1.5 thread forming tap with a spiral point for easy chip removal. It can cut threads up to 25mm deep and has a tolerance class of 6H. The tap has a straight flute design and is made in Germany by WIDIA, a leading tap manufacturer. This is a high quality thread forming tap well-suited for ductile materials.
Catalog No. 50-276-4763 Supplier Grainger Supplier No. 170FV1
The WIDIA GTD Thread Forming Tap is a high speed steel tap designed for forming threads in ductile materials like aluminum, brass, mild steels, and some plastics. It has a TiN coating for increased wear resistance and lubricity. This is a M10 x 1.5 thread forming tap with a spiral point for easy chip removal. It can cut threads up to 25mm deep and has a tolerance class of 6H. The tap has a straight flute design and is made in Germany by WIDIA, a leading tap manufacturer. This is a high quality thread forming tap well-suited for ductile materials.