Learn More
Grainger Cut-Off GDM 2020R003PF15D PR1225 10 pcs.
Catalog No. 502768563
Encompass_Preferred
The Grainger 170FN5 is a 5-gallon plastic pail with lid that is made in the USA. It features a wire handle for easy carrying and a tight sealing lid to keep contents protected. The pail is constructed from high density polyethylene and has a maximum temperature resistance of 120°F. It meets FDA regulations for food contact. The pail has approximate dimensions of 12" diameter x 11" high and weighs 2.2 lbs. This general purpose pail is ideal for storing and transporting a variety of items.
Catalog No. 50-276-8563 Supplier Grainger Supplier No. 170FN5
The Grainger 170FN5 is a 5-gallon plastic pail with lid that is made in the USA. It features a wire handle for easy carrying and a tight sealing lid to keep contents protected. The pail is constructed from high density polyethylene and has a maximum temperature resistance of 120°F. It meets FDA regulations for food contact. The pail has approximate dimensions of 12" diameter x 11" high and weighs 2.2 lbs. This general purpose pail is ideal for storing and transporting a variety of items.