Learn More
Grainger Cut-Off GDM 2020R020PM6D PR1225 10 pcs.
Catalog No. 502768586
Encompass_Preferred
The Grainger 170FC0 is a Dayton 1/3 HP Centrifugal Pump with a cast iron housing, thermoplastic impeller, and mechanical seal. It has a 115V motor with a 1.25" NPT suction and 1" NPT discharge. This centrifugal pump can deliver up to 22 GPM at 15' of head and is designed for clear water applications. The pump end materials make it suitable for pumping water, oil and mild chemicals.
Catalog No. 50-276-8586 Supplier Grainger Supplier No. 170FC0
The Grainger 170FC0 is a Dayton 1/3 HP Centrifugal Pump with a cast iron housing, thermoplastic impeller, and mechanical seal. It has a 115V motor with a 1.25" NPT suction and 1" NPT discharge. This centrifugal pump can deliver up to 22 GPM at 15' of head and is designed for clear water applications. The pump end materials make it suitable for pumping water, oil and mild chemicals.