Learn More
Grainger Cut-Off GDM 2520L003PF15D PR1535 10 pcs.
Catalog No. 502765449
Encompass_Preferred
The Grainger 170FT3 is a portable air compressor that provides 170 CFM at 100 PSI with a 5 HP electric motor. It has a 80 gallon vertical air receiver tank constructed of ASME certified steel with a durable powder coat finish. The compressor pump is oil-lubricated with a cast iron cylinder and is equipped with an automatic start/stop control to maintain tank pressure between 90-110 PSI. Safety features include an ASME safety relief valve, discharge check valve, and thermal overload protection. This air compressor is designed for industrial applications requiring high airflow.
Catalog No. 50-276-5449 Supplier Grainger Supplier No. 170FT3
The Grainger 170FT3 is a portable air compressor that provides 170 CFM at 100 PSI with a 5 HP electric motor. It has a 80 gallon vertical air receiver tank constructed of ASME certified steel with a durable powder coat finish. The compressor pump is oil-lubricated with a cast iron cylinder and is equipped with an automatic start/stop control to maintain tank pressure between 90-110 PSI. Safety features include an ASME safety relief valve, discharge check valve, and thermal overload protection. This air compressor is designed for industrial applications requiring high airflow.