Learn More
Grainger Cut-Off GDM 2520N003PF PR1535 10 pcs.
Catalog No. 502765266
Encompass_Preferred
The Grainger 170FR7 is a 1/4 HP, 115V, 60Hz, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Single Phase, Permanent Split Capacitor Electric Motor. It has a 1/4" diameter shaft and measures approximately 5-1/2" in length. The motor is thermally protected and rated for continuous duty applications. It can be used for driving fans, blowers, pumps, and other light industrial equipment. The motor is CSA certified and backed by a 1-year warranty from Grainger.
Catalog No. 50-276-5266 Supplier Grainger Supplier No. 170FR7
The Grainger 170FR7 is a 1/4 HP, 115V, 60Hz, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Single Phase, Permanent Split Capacitor Electric Motor. It has a 1/4" diameter shaft and measures approximately 5-1/2" in length. The motor is thermally protected and rated for continuous duty applications. It can be used for driving fans, blowers, pumps, and other light industrial equipment. The motor is CSA certified and backed by a 1-year warranty from Grainger.