Learn More
Grainger Cut-Off GDM 2520N010PQ PR1225 10 pcs.
Catalog No. 502765424
Encompass_Preferred
The Grainger 170FP2 is a high-quality, heavy-duty, industrial-grade, 2-ton floor jack made by Torin. It has a lift range of 5-1/2" to 18-1/8" and a lowered height of just 3-3/4". The jack is designed for use with cars, trucks, and SUVs. Key features include a steel frame, reinforced lift arm, and extra-long neck for reaching low vehicles. The jack has rear swivel casters and two front wheels for easy maneuverability. It is rated for 4,000 lb. capacity. This professional-grade floor jack is ideal for home mechanics and auto shops.
Catalog No. 50-276-5424 Supplier Grainger Supplier No. 170FP2
The Grainger 170FP2 is a high-quality, heavy-duty, industrial-grade, 2-ton floor jack made by Torin. It has a lift range of 5-1/2" to 18-1/8" and a lowered height of just 3-3/4". The jack is designed for use with cars, trucks, and SUVs. Key features include a steel frame, reinforced lift arm, and extra-long neck for reaching low vehicles. The jack has rear swivel casters and two front wheels for easy maneuverability. It is rated for 4,000 lb. capacity. This professional-grade floor jack is ideal for home mechanics and auto shops.