Learn More
Grainger Cut-Off GDM 2520N015PF PR1225 10 pcs.
Catalog No. 502764678
Encompass_Preferred
The 170FW1 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Foot Mounted Fan Motor by Century. It has a 1/2" diameter shaft and measures 5-1/2" long by 5-1/2" wide by 6-1/4" high. The motor is thermally protected and suitable for continuous duty. It can be used with air handling units, cooling towers, condensers, evaporative coolers, and more. This Century electric motor is designed for quiet, efficient operation.
Catalog No. 50-276-4678 Supplier Grainger Supplier No. 170FW1
The 170FW1 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Foot Mounted Fan Motor by Century. It has a 1/2" diameter shaft and measures 5-1/2" long by 5-1/2" wide by 6-1/4" high. The motor is thermally protected and suitable for continuous duty. It can be used with air handling units, cooling towers, condensers, evaporative coolers, and more. This Century electric motor is designed for quiet, efficient operation.