Learn More
Grainger Cut-Off GDM 2520N015PF PR1535 10 pcs.
Catalog No. 502764803
Encompass_Preferred
The Grainger 170FW7 is a Westward 3/8-inch drive flex head ratchet. It has a 72 tooth gear with a 4 degree arc swing. The head pivots in all directions for access in tight spaces. It has a chrome vanadium steel construction with a satin finish. The overall length is 10-1/2 inches with a flex head length of 2-1/2 inches. This ratchet is designed for automotive and machinery applications where access is limited. It has a lifetime warranty.
Catalog No. 50-276-4803 Supplier Grainger Supplier No. 170FW7
The Grainger 170FW7 is a Westward 3/8-inch drive flex head ratchet. It has a 72 tooth gear with a 4 degree arc swing. The head pivots in all directions for access in tight spaces. It has a chrome vanadium steel construction with a satin finish. The overall length is 10-1/2 inches with a flex head length of 2-1/2 inches. This ratchet is designed for automotive and machinery applications where access is limited. It has a lifetime warranty.