Learn More
Grainger Cut-Off GDM 2520N020PM PR1535 10 pcs.
Catalog No. 502764736
Encompass_Preferred
The 170FV2 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor from Marathon Motors. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is designed for general purpose applications and features a cast iron frame and end bells along with ball bearings for smooth, reliable operation. With a continuous duty rating, it can run at full load for extended periods. This electric motor would be well-suited for powering pumps, fans, conveyors, compressors, machine tools, and other industrial equipment.
Catalog No. 50-276-4736 Supplier Grainger Supplier No. 170FV2
The 170FV2 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor from Marathon Motors. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is designed for general purpose applications and features a cast iron frame and end bells along with ball bearings for smooth, reliable operation. With a continuous duty rating, it can run at full load for extended periods. This electric motor would be well-suited for powering pumps, fans, conveyors, compressors, machine tools, and other industrial equipment.