Learn More
Grainger Cut-Off GDM 3020N003PF PR1225 10 pcs.
Catalog No. 502765776
Encompass_Preferred
The Grainger 170FN7 is a 1/4 HP, 115V, 60Hz, 1 phase, 3450 RPM, 56C frame, open drip proof, continuous duty, C-face motor. It has a 48Y frame with a 1.25" shaft diameter and 1.38" shaft height. The motor is thermally protected and rated for indoor use. It has ball bearings and measures approximately 9-1/2" in length by 5-3/4" in width by 6-7/8" in height. The motor weighs approximately 18 pounds.
Catalog No. 50-276-5776 Supplier Grainger Supplier No. 170FN7
Add to Cart
Add to Cart
The Grainger 170FN7 is a 1/4 HP, 115V, 60Hz, 1 phase, 3450 RPM, 56C frame, open drip proof, continuous duty, C-face motor. It has a 48Y frame with a 1.25" shaft diameter and 1.38" shaft height. The motor is thermally protected and rated for indoor use. It has ball bearings and measures approximately 9-1/2" in length by 5-3/4" in width by 6-7/8" in height. The motor weighs approximately 18 pounds.