Learn More
Grainger Cut-Off GDM 3020N015PF PR1225 10 pcs.
Catalog No. 502764761
Encompass_Preferred
The Grainger 170FW2 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof Fan Motor. It has a 1/3 horsepower and operates on 115/230 volt AC power at 1725 RPM. The motor has a 56C frame and an open drip proof enclosure. This electric motor is designed for circulating air or exhaust applications and is suitable for use with fans or blowers.
Catalog No. 50-276-4761 Supplier Grainger Supplier No. 170FW2
Add to Cart
Add to Cart
The Grainger 170FW2 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof Fan Motor. It has a 1/3 horsepower and operates on 115/230 volt AC power at 1725 RPM. The motor has a 56C frame and an open drip proof enclosure. This electric motor is designed for circulating air or exhaust applications and is suitable for use with fans or blowers.