Learn More
Grainger Cut-Off GDM 3020N015PF PR1535 10 pcs.
Catalog No. 502764804
Encompass_Preferred
The Grainger 170FW8 is a 1/2 HP, 115V, single phase, open drip proof fan motor with a 1075 RPM operating speed. It features a 48 frame size, CCW rotation (when viewed from shaft end), and includes permanently lubricated sleeve bearings. This motor is thermally protected and suitable for continuous duty applications. With its open ventilated design, it is well suited for use in industrial and commercial ventilation systems. The motor dimensions are 6-1/2" diameter x 8-3/8" long.
Catalog No. 50-276-4804 Supplier Grainger Supplier No. 170FW8
The Grainger 170FW8 is a 1/2 HP, 115V, single phase, open drip proof fan motor with a 1075 RPM operating speed. It features a 48 frame size, CCW rotation (when viewed from shaft end), and includes permanently lubricated sleeve bearings. This motor is thermally protected and suitable for continuous duty applications. With its open ventilated design, it is well suited for use in industrial and commercial ventilation systems. The motor dimensions are 6-1/2" diameter x 8-3/8" long.