Learn More
Grainger Cut-Off GDM 3020R010PQ15D PR1535 10 pcs.
Catalog No. 502764805
Encompass_Preferred
The Grainger 170FV0 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and ball bearings. It can be used for general purpose applications requiring 1/2 HP.
Catalog No. 50-276-4805 Supplier Grainger Supplier No. 170FV0
The Grainger 170FV0 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and ball bearings. It can be used for general purpose applications requiring 1/2 HP.