Learn More
Grainger Cut-Off GDM 3020R015PF15D PR1535 10 pcs.
Catalog No. 502764840
Encompass_Preferred
The Grainger 170FX0 is a high-quality, durable, and versatile air compressor. This 2 HP, oil-lubricated, twin tank compressor has a maximum pressure of 175 PSI and delivers 14.5 CFM at 90 PSI. It features a cast iron, V-twin cylinder pump for long life and quiet operation. The compressor includes an ASME coded tank, thermal overload protection, and an oil sight glass. This compressor is ideal for automotive service, construction, manufacturing, woodworking shops, auto body shops, and anywhere industrial-quality compressed air is needed. The twin tank design provides reduced pump cycling for longer life.
Catalog No. 50-276-4840 Supplier Grainger Supplier No. 170FX0
The Grainger 170FX0 is a high-quality, durable, and versatile air compressor. This 2 HP, oil-lubricated, twin tank compressor has a maximum pressure of 175 PSI and delivers 14.5 CFM at 90 PSI. It features a cast iron, V-twin cylinder pump for long life and quiet operation. The compressor includes an ASME coded tank, thermal overload protection, and an oil sight glass. This compressor is ideal for automotive service, construction, manufacturing, woodworking shops, auto body shops, and anywhere industrial-quality compressed air is needed. The twin tank design provides reduced pump cycling for longer life.