Learn More
Grainger Cut-Off GDM 3020R025PM6D PR1225 10 pcs.
Catalog No. 502765834
Encompass_Preferred
The Grainger 170FC4 is a Dayton 1/3 HP Centrifugal Pump with a cast iron housing, thermoplastic impeller, and mechanical seal. It has a 115V motor with a 1.25" NPT suction and 1" NPT discharge. This centrifugal pump can deliver up to 22 GPM at 15' of head and is designed for clear water applications. The pump end materials make it suitable for pumping water, oil and mild chemicals.
Catalog No. 50-276-5834 Supplier Grainger Supplier No. 170FC4
Add to Cart
Add to Cart
The Grainger 170FC4 is a Dayton 1/3 HP Centrifugal Pump with a cast iron housing, thermoplastic impeller, and mechanical seal. It has a 115V motor with a 1.25" NPT suction and 1" NPT discharge. This centrifugal pump can deliver up to 22 GPM at 15' of head and is designed for clear water applications. The pump end materials make it suitable for pumping water, oil and mild chemicals.