Learn More
Grainger Cut-Off GDM 4020N030PM PR1225 10 pcs.
Catalog No. 502765323
Encompass_Preferred
The Grainger 170FA8 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It has a 48Y frame with a continuous duty rating and manual overload protector. The motor is totally enclosed, fan cooled (TEFC) with ball bearings and a C-face mounting. It can be used for driving fans, blowers, pumps, conveyors, and other industrial machinery. The motor dimensions are 5-7/8" diameter x 7-3/8" long with a 1/2" diameter shaft. It weighs approximately 13 pounds.
Catalog No. 50-276-5323 Supplier Grainger Supplier No. 170FA8
The Grainger 170FA8 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It has a 48Y frame with a continuous duty rating and manual overload protector. The motor is totally enclosed, fan cooled (TEFC) with ball bearings and a C-face mounting. It can be used for driving fans, blowers, pumps, conveyors, and other industrial machinery. The motor dimensions are 5-7/8" diameter x 7-3/8" long with a 1/2" diameter shaft. It weighs approximately 13 pounds.