Learn More
Grainger Cut-Off GDM 4020N030PM PR1535 10 pcs.
Catalog No. 502764361
Encompass_Preferred
The 170FV4 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor manufactured by Baldor-Reliance. It has a 1.15 service factor, CCW rotation when viewed from the shaft end, and features a cast iron frame, cast iron junction box, and cast iron end brackets. The motor is totally enclosed, fan cooled (TEFC) with a class F insulation and NEMA standard measurements.
Catalog No. 50-276-4361 Supplier Grainger Supplier No. 170FV4
Add to Cart
Add to Cart
The 170FV4 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor manufactured by Baldor-Reliance. It has a 1.15 service factor, CCW rotation when viewed from the shaft end, and features a cast iron frame, cast iron junction box, and cast iron end brackets. The motor is totally enclosed, fan cooled (TEFC) with a class F insulation and NEMA standard measurements.