Learn More
Grainger Cut-Off TK R3 CR9025 CVD C 10 pcs.
Catalog No. 502766144
Encompass_Preferred
The 170EU4 is a 3M Novec 1230 Fire Suppression System Cylinder, 150 lb capacity, designed for total flooding applications. It uses an environmentally sustainable clean agent that is electrically non-conductive and leaves no residue. This cylinder has a working pressure of 360 psi at 70°F and is DOT 4BW500 rated. It can be used with the 3M Novec 1230 fire suppression system to protect areas like telecommunications facilities, archives, and data centers. The cylinder is made of aluminum for light weight and corrosion resistance.
Catalog No. 50-276-6144 Supplier Grainger Supplier No. 170EU4
The 170EU4 is a 3M Novec 1230 Fire Suppression System Cylinder, 150 lb capacity, designed for total flooding applications. It uses an environmentally sustainable clean agent that is electrically non-conductive and leaves no residue. This cylinder has a working pressure of 360 psi at 70°F and is DOT 4BW500 rated. It can be used with the 3M Novec 1230 fire suppression system to protect areas like telecommunications facilities, archives, and data centers. The cylinder is made of aluminum for light weight and corrosion resistance.