Learn More
Grainger Cut-Off TK R3 KW10 Uncoate 10 pcs.
Catalog No. 502765281
Encompass_Preferred
The 170EU5 is a 5-gallon plastic utility pail manufactured by Rubbermaid Commercial Products. It is made from high-density polyethylene and has a wire bail handle for easy lifting and carrying. The pail has graduated measurements on the side for accurate liquid measurement. It can hold up to 40 lbs and has a tapered shape for easy stacking. The pail is ideal for commercial and industrial uses such as mixing paints, transporting liquids, storing bulk materials, and more. It provides a durable and economical container solution for a variety of applications.
Catalog No. 50-276-5281 Supplier Grainger Supplier No. 170EU5
The 170EU5 is a 5-gallon plastic utility pail manufactured by Rubbermaid Commercial Products. It is made from high-density polyethylene and has a wire bail handle for easy lifting and carrying. The pail has graduated measurements on the side for accurate liquid measurement. It can hold up to 40 lbs and has a tapered shape for easy stacking. The pail is ideal for commercial and industrial uses such as mixing paints, transporting liquids, storing bulk materials, and more. It provides a durable and economical container solution for a variety of applications.