Learn More
Grainger Cut-Off TK R3P KW10 Uncoat 10 pcs.
Catalog No. 502765790
Encompass_Preferred
The 170EV3 is a 3-phase, 460V AC, 60Hz, 1700 RPM, 5 HP electric motor manufactured by Baldor-Reliance. It is an inverter duty motor with a 1.15 service factor, TEFC enclosure, regreaseable bearings, and meets NEMA premium efficiency standards. The motor has a C-face mounting, 182T frame size, and features a cast iron construction with Class F insulation. It can provide continuous duty operation for industrial applications requiring reliable 3-phase power.
Catalog No. 50-276-5790 Supplier Grainger Supplier No. 170EV3
The 170EV3 is a 3-phase, 460V AC, 60Hz, 1700 RPM, 5 HP electric motor manufactured by Baldor-Reliance. It is an inverter duty motor with a 1.15 service factor, TEFC enclosure, regreaseable bearings, and meets NEMA premium efficiency standards. The motor has a C-face mounting, 182T frame size, and features a cast iron construction with Class F insulation. It can provide continuous duty operation for industrial applications requiring reliable 3-phase power.