Learn More
Grainger Cut-Off TK R4 CR9025 CVD C 10 pcs.
Catalog No. 502765833
Encompass_Preferred
The Grainger 170EU7 is a Westinghouse 7.5 HP, 230/460V, 1725 RPM, 3 phase, open drip proof electric motor. It has a cast iron frame, thermally protected, and is rated for continuous duty. The motor has a NEMA design B with a standard foot mounting, C-face endshields, and 1.15 service factor. It can provide up to 5.6 kW of mechanical power. This electric motor would be well suited for industrial applications requiring reliable 3 phase power.
Catalog No. 50-276-5833 Supplier Grainger Supplier No. 170EU7
The Grainger 170EU7 is a Westinghouse 7.5 HP, 230/460V, 1725 RPM, 3 phase, open drip proof electric motor. It has a cast iron frame, thermally protected, and is rated for continuous duty. The motor has a NEMA design B with a standard foot mounting, C-face endshields, and 1.15 service factor. It can provide up to 5.6 kW of mechanical power. This electric motor would be well suited for industrial applications requiring reliable 3 phase power.