Learn More
Grainger Cut-Off TKF 12L050NB20DR KW10 10 pcs.
Catalog No. 502765493
Encompass_Preferred
The Grainger 170EW7 is a Westinghouse 7.5 HP, 230/460V, 1725 RPM, 3 Phase, TEFC Electric Motor. It features a cast iron frame, thermally protected, 1.15 service factor, and standard foot mounting. This electric motor is suitable for industrial applications requiring reliable 3 phase power.
Catalog No. 50-276-5493 Supplier Grainger Supplier No. 170EW7
Add to Cart
Add to Cart
The Grainger 170EW7 is a Westinghouse 7.5 HP, 230/460V, 1725 RPM, 3 Phase, TEFC Electric Motor. It features a cast iron frame, thermally protected, 1.15 service factor, and standard foot mounting. This electric motor is suitable for industrial applications requiring reliable 3 phase power.